Cleanroom overalls, BioClean-D™
Leverancier: Nitritex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S-BDFC-XL S-BDFC-L S-BDFC-XXL S-BDFC-XXXL S-BDFC-S S-BDFC-XS S-BDFC-M S-BDFC-XXXXL BDFC-L BDFC-XL BDFC-XXL
NITRS-BDFC-XLEA 380 EUR
NITRS-BDFC-XL NITRS-BDFC-L NITRS-BDFC-XXL NITRS-BDFC-XXXL NITRS-BDFC-S NITRS-BDFC-XS NITRS-BDFC-M NITRS-BDFC-XXXXL NITRBDFC-L NITRBDFC-XL NITRBDFC-XXL
Cleanroom overalls, BioClean-D™
Wegwerpkleding
The BioClean-D overall with hood and integrated boots is a disposable garment offering comfort and head-to-toe protection.
  • Light weight and low-linting
  • CleanTough material
  • Thumb loops to ensure a secure hold
  • Zip with sealable cover
  • Elasticated hood, back, cuffs and ankles
  • Slip-resistant soles
The BioClean-D overall with hood and integrated boots features a front zip closure with protective flap, elasticated hood, back, cuffs and ankles, and thumb loops to ensure a secure hold. The integrated boots feature slip-resistant soles to ensure every step isMore Product Information