Anti-RAD50 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-154 NB100-154SS
NOVUNB100-154EA 624 EUR
NOVUNB100-154 NOVUNB100-154SS
Anti-RAD50 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: RAD50 (RAD50 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR