Anti-CFC1 Rabbit Polyclonal Antibody (DyLight® 405)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-1598V
NOVUNB100-1598VEA 723 EUR
NOVUNB100-1598V
Anti-CFC1 Rabbit Polyclonal Antibody (DyLight® 405)
Antilichamen
Type: Primary
Antigen: CFC1 (cripto, FRL-1, cryptic family 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR