Anti-HMGB1 Rabbit Polyclonal Antibody (Biotin)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-2322B
NOVUNB100-2322BEA 792 EUR
NOVUNB100-2322B
Anti-HMGB1 Rabbit Polyclonal Antibody (Biotin)
Antilichamen
Type: Primary
Antigen: HMGB1 (high mobility group box 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR