Anti-MAP1LC3A Rabbit Polyclonal Antibody (DyLight® 488)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-2331G
NOVUNB100-2331GEA 806 EUR
NOVUNB100-2331G
Anti-MAP1LC3A Rabbit Polyclonal Antibody (DyLight® 488)
Antilichamen
Type: Primary
Antigen: MAP1LC3A (microtubule-associated protein 1 light chain 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR