Anti-VEGFA Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-2381F
NOVUNB100-2381FEA 743 EUR
NOVUNB100-2381F
Anti-VEGFA Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Specificaties

antigeen symbool VEGFA
Type antilichaam Primair,Vascular endothelial growth factor A
Environmentally Preferable
Antigeen synoniemen MVCD1|VEGF|VPF
Type antilichaam Primair,Vascular endothelial growth factor A
Clonality Polyklonaal
Conjugatie FITC (Fluoresceïne-isothiocyanaat)
Reactivity
Host Rabbit
Gen-ID 7422
Isotype
Western blot Yes
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR