Anti-S-arrestin Mouse Monoclonal Antibody (DyLight® 650) [clone: PDS-1]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-2385C
NOVUNB100-2385CEA 668 EUR
NOVUNB100-2385C
Anti-S-arrestin Mouse Monoclonal Antibody (DyLight® 650) [clone: PDS-1]
Antilichamen
Type: Primary
Antigen: S-arrestin
Clonality: monoclonal
Clone: PDS-1
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR