Anti-BUB1B Mouse Monoclonal Antibody [clone: 8G1]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-353 NB100-353SS
NOVUNB100-353EA 630 EUR
NOVUNB100-353 NOVUNB100-353SS
Anti-BUB1B Mouse Monoclonal Antibody [clone: 8G1]
Antilichamen
Type: Primary
Antigen: BUB1B (BUB1 mitotic checkpoint serine/threonine kinase B)
Clonality: monoclonal
Clone: 8G1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR