Anti-TMED4 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-56393
NOVUNB100-56393EA 443 EUR
NOVUNB100-56393
Anti-TMED4 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: TMED4 (transmembrane emp24 protein transport domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR