Anti-rRNA Mouse Monoclonal Antibody (DyLight® 550) [clone: Y10b]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-662R
NOVUNB100-662REA 806 EUR
NOVUNB100-662R
Anti-rRNA Mouse Monoclonal Antibody (DyLight® 550) [clone: Y10b]
Antilichamen
Type: Primary
Antigen: rRNA
Clonality: monoclonal
Clone: Y10b
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Pan
Order Now

Specificaties

antigeen symbool rRNA
Environmentally Preferable
Type antilichaam Primair
Clonality Monoklonaal
Conjugatie DyLight® 550
Clone Y10b
Reactivity Multiple
Host Mouse
Isotype IgG3 kappa
ImmunoNeerslag Yes
ImmunoFluorescentie Yes
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR