Anti-DISC1 Rabbit Polyclonal Antibody (Biotin)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB110-40775B
NOVUNB110-40775BEA 668 EUR
NOVUNB110-40775B
Anti-DISC1 Rabbit Polyclonal Antibody (Biotin)
Antilichamen
Type: Primary
Antigen: DISC1 (disrupted in schizophrenia 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR