Anti-LRRK2 Rabbit Polyclonal Antibody (DyLight® 405)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB110-55289V
NOVUNB110-55289VEA 668 EUR
NOVUNB110-55289V
Anti-LRRK2 Rabbit Polyclonal Antibody (DyLight® 405)
Antilichamen
Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR