Anti-ALOX5 Rabbit Polyclonal Antibody (DyLight® 488)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB110-58748G
NOVUNB110-58748GEA 668 EUR
NOVUNB110-58748G
Anti-ALOX5 Rabbit Polyclonal Antibody (DyLight® 488)
Antilichamen
Type: Primary
Antigen: ALOX5 (arachidonate 5-lipoxygenase)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR