Anti-BECN2 Rabbit Polyclonal Antibody (DyLight® 550)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB110-60982R
NOVUNB110-60982REA 668 EUR
NOVUNB110-60982R
Anti-BECN2 Rabbit Polyclonal Antibody (DyLight® 550)
Antilichamen
Type: Primary
Antigen: BECN2 (beclin 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR