Anti-PIK3C3 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB110-87320 NB110-87320SS
NOVUNB110-87320EA 622 EUR
NOVUNB110-87320 NOVUNB110-87320SS
Anti-PIK3C3 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: PIK3C3 (phosphatidylinositol 3-kinase, catalytic subunit type 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR