Anti-TRDMT1 Mouse Monoclonal Antibody [clone: 102B1259.2]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB200-587
NOVUNB200-587EA 558 EUR
NOVUNB200-587
Anti-TRDMT1 Mouse Monoclonal Antibody [clone: 102B1259.2]
Antilichamen
Type: Primary
Antigen: TRDMT1 (tRNA aspartic acid methyltransferase 1)
Clonality: monoclonal
Clone: 102B1259.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR