Anti-HIST3H3 Rabbit Polyclonal Antibody (DyLight® 650)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB21-1092C
NOVUNB21-1092CEA 836 EUR
NOVUNB21-1092C
Anti-HIST3H3 Rabbit Polyclonal Antibody (DyLight® 650)
Antilichamen
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR