Anti-PSENEN Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB300-227 NB300-227SS
NOVUNB300-227EA 568 EUR
NOVUNB300-227 NOVUNB300-227SS
Anti-PSENEN Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: PSENEN (presenilin enhancer gamma secretase subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR