Anti-FXR2 Mouse Monoclonal Antibody (DyLight® 488) [clone: 1G2]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB300-259G
NOVUNB300-259GEA 655 EUR
NOVUNB300-259G
Anti-FXR2 Mouse Monoclonal Antibody (DyLight® 488) [clone: 1G2]
Antilichamen
Type: Primary
Antigen: FXR2 (fragile X mental retardation, autosomal homolog 2)
Clonality: monoclonal
Clone: 1G2
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR