Anti-NES Mouse Monoclonal Antibody (DyLight® 650) [clone: 10C2]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB300-266C
NOVUNB300-266CEA 736 EUR
NOVUNB300-266C
Anti-NES Mouse Monoclonal Antibody (DyLight® 650) [clone: 10C2]
Antilichamen
Type: Primary
Antigen: NES (nestin)
Clonality: monoclonal
Clone: 10C2
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR