Anti-SCARB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB400-104PE
NOVUNB400-104PEEA 751 EUR
NOVUNB400-104PE
Anti-SCARB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antilichamen
Type: Primary
Antigen: SCARB1 (scavenger receptor class B, member 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR