Anti-THOP1 Mouse Monoclonal Antibody [clone: 4D6]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB400-146 NB400-146SS
NOVUNB400-146EA 554 EUR
NOVUNB400-146 NOVUNB400-146SS
Anti-THOP1 Mouse Monoclonal Antibody [clone: 4D6]
Antilichamen
The Thimet Oligopeptidase / THOP1 Antibody (4D6) from Novus Biologicals is a mouse monoclonal antibody to Thimet Oligopeptidase / THOP1. This antibody reacts with human, mouse, rat, primate, rabbit. The Thimet Oligopeptidase / THOP1 Antibody (4D6) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: THOP1
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
More Product Information
Order Now

Specificaties

antigeen symbool THOP1
Environmentally Preferable
Type antilichaam Primair,Thimet Oligopeptidase 1
Antigeen synoniemen MP78|TOP|MEPD_HUMAN|EP24.15
Type antilichaam Primair,Thimet Oligopeptidase 1
Clonality Monoklonaal
Conjugatie Onconjugeerd
Clone 4D6
Reactivity Human
Host Mouse
Gen-ID 7064
Isotype IgG2b
Western blot Yes
ImmunoFluorescentie Yes
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR