Anti-POT1 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB500-176 NB500-176SS
NOVUNB500-176EA 642 EUR
NOVUNB500-176 NOVUNB500-176SS
Anti-POT1 Rabbit Polyclonal Antibody
Antilichamen
The POT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to POT1. This antibody reacts with human. The POT1 Antibody has been validated for the following applications: Western Blot.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: POT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR