Anti-NCL Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB600-241 NB600-241SS
NOVUNB600-241EA 588 EUR
NOVUNB600-241 NOVUNB600-241SS
Anti-NCL Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: NCL (nucleolin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR