Anti-MYC Mouse Monoclonal Antibody (DyLight® 755) [clone: 9E10]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB600-302IR
NOVUNB600-302IREA 710 EUR
NOVUNB600-302IR
Anti-MYC Mouse Monoclonal Antibody (DyLight® 755) [clone: 9E10]
Antilichamen
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: 90000000000
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR