Anti-MYC Rabbit Polyclonal Antibody (DyLight® 488)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB600-336G
NOVUNB600-336GEA 743 EUR
NOVUNB600-336G
Anti-MYC Rabbit Polyclonal Antibody (DyLight® 488)
Antilichamen
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR