Anti-HPd1 Mouse Monoclonal Antibody (DyLight® 755) [clone: DHIC2-4A10]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-18953IR
NOVUNBP1-18953IREA 668 EUR
NOVUNBP1-18953IR
Anti-HPd1 Mouse Monoclonal Antibody (DyLight® 755) [clone: DHIC2-4A10]
Antilichamen
Type: Primary
Antigen: HPd1
Clonality: monoclonal
Clone: DHIC2-4A10
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR