Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 680) [clone: OC2-3C7]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-18970FR
NOVUNBP1-18970FREA 668 EUR
NOVUNBP1-18970FR
Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 680) [clone: OC2-3C7]
Antilichamen
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-3C7
Conjugation: DyLight® 680
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR