Anti-Palladin Mouse Monoclonal Antibody (DyLight® 680) [clone: 1E6]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-25959FR
NOVUNBP1-25959FREA 722 EUR
NOVUNBP1-25959FR
Anti-Palladin Mouse Monoclonal Antibody (DyLight® 680) [clone: 1E6]
Antilichamen
Type: Primary
Antigen: Palladin
Clonality: monoclonal
Clone: 1000000
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR