Anti-HIF1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: exon 12]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-47180H
NOVUNBP1-47180HEA 722 EUR
NOVUNBP1-47180H
Anti-HIF1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: exon 12]
Antilichamen
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone: exon 12
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Specificaties

antigeen symbool HIF1A
Environmentally Preferable
Type antilichaam Primair,HIF1 alpha
Antigeen synoniemen PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
Type antilichaam Primair,HIF1 alpha
Clonality Polyklonaal
Conjugatie HRP (Horseradish Peroxidase)
Clone exon 12
Reactivity Human
Host Rabbit
Gen-ID 3091
Isotype
Western blot Yes
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR