Anti-SLITRK1 Rabbit Polyclonal Antibody (DyLight® 650)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-49541C
NOVUNBP1-49541CEA 668 EUR
NOVUNBP1-49541C
Anti-SLITRK1 Rabbit Polyclonal Antibody (DyLight® 650)
Antilichamen
Type: Primary
Antigen: SLITRK1 (SLIT and NTRK-like family, member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR