Anti-AURKA Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-51843 NBP1-51843SS
NOVUNBP1-51843EA 606 EUR
NOVUNBP1-51843 NOVUNBP1-51843SS
Anti-AURKA Rabbit Polyclonal Antibody
Antilichamen
The Aurora A Antibody from Novus Biologicals is a rabbit polyclonal antibody to Aurora A. This antibody reacts with human, mouse. The Aurora A Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Simple Western

Type: Primary
Antigen: Aurora A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Specificaties

antigeen symbool AURKA
Environmentally Preferable
Type antilichaam Primair,Aurora 2
Antigeen synoniemen BTAK|AURA|STK7|ARK1|PPP1R47|Serine/threonine-protein kinase 12|STK6|AURORA2|Aurora-B|STK15|AIK
Type antilichaam Primair,Aurora 2
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Gen-ID 6790
Isotype
Western blot Yes
ImmunoFluorescentie Yes
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR