Anti-TEK Rabbit Polyclonal Antibody (Biotin)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-69753B
NOVUNBP1-69753BEA 722 EUR
NOVUNBP1-69753B
Anti-TEK Rabbit Polyclonal Antibody (Biotin)
Antilichamen
Type: Primary
Antigen: TEK (TEK tyrosine kinase, endothelial)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR