Anti-PTCH1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-71662H
NOVUNBP1-71662HEA 722 EUR
NOVUNBP1-71662H
Anti-PTCH1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
Type: Primary
Antigen: PTCH1 (patched 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR