Anti-GABARAP Rabbit Polyclonal Antibody (DyLight® 550)

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-71771R
NOVUNBP1-71771REA 668 EUR
NOVUNBP1-71771R
Anti-GABARAP Rabbit Polyclonal Antibody (DyLight® 550)
Antilichamen
Type: Primary
Antigen: GABARAP (GABA(A) receptor-associated protein)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR