Anti-DSG3 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5H10]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-78984F
NOVUNBP1-78984FEA 668 EUR
NOVUNBP1-78984F
Anti-DSG3 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5H10]
Antilichamen
Type: Primary
Antigen: DSG3 (desmoglein 3)
Clonality: monoclonal
Clone: 5H10
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR