Anti-S100A9 Rabbit Polyclonal Antibody
Leverancier: Novus Biologicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
NBP1-89360-25
NOVUNBP1-89360-25EA 443 EUR
NOVUNBP1-89360-25
Anti-S100A9 Rabbit Polyclonal Antibody
Antilichamen
S100A9 is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localised in the cytoplasm and/or nucleus of a wide range of cells, and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100A9 may function in the inhibition of casein kinase, and altered expression is associated with the disease cystic fibrosis. Overexpression of S100A9 has also recently been linked to a poor prognosis with invasive ductal carcinoma of the breast.
Opgelet: For research use only.