Anti-GOLGB1 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-91938
NOVUNBP1-91938EA 635 EUR
NOVUNBP1-91938
Anti-GOLGB1 Rabbit Polyclonal Antibody
Antilichamen
The GOLGB1 / Giantin Antibody from Novus Biologicals is a rabbit polyclonal antibody to GOLGB1 / Giantin. This antibody reacts with human. The GOLGB1 / Giantin Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: GOLGB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR