Anti-HDAC7 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP2-03979
NOVUNBP2-03979EA 494 EUR
NOVUNBP2-03979
Anti-HDAC7 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: HDAC7 (histone deacetylase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Specificaties

antigeen symbool HDAC7
Type antilichaam Primair,Histone deacetylase 7
Environmentally Preferable
Antigeen synoniemen HD7A|HD7|HDAC7A
Type antilichaam Primair,Histone deacetylase 7
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Gen-ID 51564
Isotype
Western blot Yes
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR