Anti-MMP9 Mouse Monoclonal Antibody (Biotin) [clone: 4A3]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP2-13173B
NOVUNBP2-13173BEA 703 EUR
NOVUNBP2-13173B
Anti-MMP9 Mouse Monoclonal Antibody (Biotin) [clone: 4A3]
Antilichamen
Type: Primary
Antigen: MMP9 (matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase))
Clonality: monoclonal
Clone: 4A3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR