Anti-RUNX2 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP2-24755
NOVUNBP2-24755EA 558 EUR
NOVUNBP2-24755
Anti-RUNX2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: RUNX2 (runt-related transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR