Anti-TLR6 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP2-24914
NOVUNBP2-24914EA 434 EUR
NOVUNBP2-24914
Anti-TLR6 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: TLR6 (toll-like receptor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR