Anti-FOXP3 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 3G3]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP2-26668
NOVUNBP2-26668EA 704 EUR
NOVUNBP2-26668
Anti-FOXP3 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 3G3]
Antilichamen
Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: monoclonal
Clone: 3G3
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR