Anti-CD138 Mouse Monoclonal Antibody [clone: 1A3H4]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP2-37282
NOVUNBP2-37282EA 675 EUR
NOVUNBP2-37282
Anti-CD138 Mouse Monoclonal Antibody [clone: 1A3H4]
Antilichamen
This antibody is CyTOF-ready.

  • ELISA 1:10000
  • Flow Cytometry 1:200 - 1:400
  • Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000
  • Immunohistochemistry 1:200 - 1:1000
  • Immunohistochemistry-Paraffin 1:200 - 1:1000
  • Western Blot 1:500 - 1:2000

this antibody is reactive to Mouse and derived from the same host, Mouse. Mouse-On-Mouse blocking reagent may be needed for IHC and ICC experiments to reduce high background signal.

Opgelet: This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Type: Primary
Antigen:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR