Standard opening screw thread vials
Leverancier: THERMO NATIONAL SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C4013-1 C4013-2 C4013-11
NSCAC4013-1EA 45.5 EUR
NSCAC4013-1 NSCAC4013-2 NSCAC4013-11
Standard opening screw thread vials
flesjes
Preferred for use with many carousel tray autosamplers. Glass vials are manufactured from superior quality borosilicate clear glass Type 1.
  • Re-usable sleeves
Microsampling and high recovery vials allow maximum sample extraction without need for separate inserts.