Supelco® Organic Reference Standard, F.A.M.E. Mix RM-1

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

O7006-1AMP
O7006-1AMPEA 134 EUR
O7006-1AMP
Supelco® Organic Reference Standard, F.A.M.E. Mix RM-1
Standaarden Organische standaarden

Palmitic acid methyl ester (C16:0) 6,0% Stearic acid methyl ester (C18:0) 3,0% Oleic acid methyl ester (C18:1cis-9) 35,0% Linoleic acid methyl ester (C18:2cis-912) 50,0% Linolenic acid methyl ester (C18:3cis-91215) 3,0% Arachidic acid methyl ester (C20:0) 3,0%.


Opgelet: P273 - P501

Order Now

Specificaties

Synonym(s) AOCS Reference Mixtures; AOCS No. 1 (RM-1); Oil Reference Standard, AOCS No. 1; Oil Reference Standard, AOCS
Environmentally Preferable
M-Clarity quality level MQ100
Beschrijving van de gevarenklasse H413
Shipping conditions DICE
Storage conditions –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR