Rocking shakers, analogue
Leverancier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30391961
OHAU30391961EA 1140 EUR
OHAU30391961
Rocking shakers, analogue
Schudapparaten Roterende/schommelende schudapparaten
These analogue rocking shakers are easy and economical options for all the rocking needs in laboratory. Ideal for cell culture and blotting applications and also designed to be used in a variety of environmental conditions. The aluminium base offers durability and added stability.
  • Takes up less space and easily fits into most hoods and incubators
  • Microprocessor control provides tilt adjustment which allows user to easily adjust waving angle from 0 to 15° while unit is operating
  • Independent control knobs for speed, tilt, and time, allow for easy adjustments
More Product Information