Accessories for platforms and adjustable bars for analogue/digital and reciprocating shakers
Leverancier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30400052 30400053 30400054 30400056 30400057 30400058 30400059 30400066 30400067 30400068 30400069 30400070 30400071 30400072 30400073 30400074 30400075 30400076 30400077 30400078 30400079 30400080 30400081 30400082 30400083 30400121 30400122
OHAU30400052EA 214 EUR
OHAU30400052 OHAU30400053 OHAU30400054 OHAU30400056 OHAU30400057 OHAU30400058 OHAU30400059 OHAU30400066 OHAU30400067 OHAU30400068 OHAU30400069 OHAU30400070 OHAU30400071 OHAU30400072 OHAU30400073 OHAU30400074 OHAU30400075 OHAU30400076 OHAU30400077 OHAU30400078 OHAU30400079 OHAU30400080 OHAU30400081 OHAU30400082 OHAU30400083 OHAU30400121 OHAU30400122
Accessories for platforms and adjustable bars for analogue/digital and reciprocating shakers
Schudapparaten Schudincubatoren
Universal platforms: Ideal for mounting flask clamps, test tube racks, and bottle clamps. The two-tier braces allow stacking of platforms with a 25,4 cm clearance (available for select sizes). An optional non-skid rubber mat can be placed on the platform for an added non-slip surface. The platform is constructed of type 304 stainless steel.
Culture platforms: Ideal for slow speed applications; Petri dishes, culture flasks, and other flat bottom, low profile vessels. The platform has a non-skid rubber surface.
Adjustable platforms: Adjustable clamping bars accommodate various vesselMore Product Information
Order Now
Specificaties
Environmentally Preferable