Trays for digital/analogue rocking and waving shakers
Leverancier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30400137 30400138 30400139
OHAU30400137EA 134 EUR
OHAU30400137 OHAU30400138 OHAU30400139
Trays for digital/analogue rocking and waving shakers
Reservoirs/Schalen/Potten Instrumentenschalen
These trays expand the capacity of the shakers by adding a second tier to the shaker.
  • Easily attaches to the unit, includes tray to add a second tier for higher capacity applications
  • Second tier tray mounts 8,9 cm above the lower tray
Leveringsinformatie: Tray includes hardware and a rubber mat.