Anti-NXNL2 Mouse Monoclonal Antibody [clone: 1C3]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CF501215
ORIGCF501215EA 833 EUR
ORIGCF501215
Anti-NXNL2 Mouse Monoclonal Antibody [clone: 1C3]
Antilichamen
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: NXNL2
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR